siRNA Details
Gene Symbol: Akr1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145364.2 | NP_663339.1 | 4, 5 | 532 | ||
GenBank mRNA | BC018333.1 | - | 4, 5 | 532 | 4162788 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Akr1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145364.2 | NP_663339.1 | 4, 5 | 532 | ||
GenBank mRNA | BC018333.1 | - | 4, 5 | 532 | 4162788 |