siRNA Details
| Gene Symbol: Stk10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009288.2 | NP_033314.2 | 11 | 1845 | ||
| GenBank mRNA | AK148118.1 | - | 11 | 1788 | ||
| AK220448.1 | - | 11 | 1767 | |||
| BC069840.1 | - | 11 | 1918 | |||
| BC096624.1 | - | 11 | 1930 | |||
| BC128363.1 | - | 11 | 1731 | 40111177 | ||
| D89728.1 | - | 11 | 1806 | |||

Stk10
Genomic Map