siRNA Details
| Gene Symbol: Stk11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB015801.1 | - | 1 | 132 | ||
| AF129870.1 | - | 1 | 510 | |||
| AF145287.1 | - | 1 | 114 | |||
| AF151711.1 | - | 1 | 114 | |||
| AK171909.1 | - | 1 | 908 | |||
| AK172385.1 | - | 1 | 897 | |||
| AK172528.1 | - | 1 | 894 | |||
| BC052379.1 | - | 1 | 175 | 5688695 | ||
| BE951985.1 | - | 1 | 377 | |||
| CO424479.1 | - | 1 | 332 | |||
| EU730638.1 | - | 1 | 93 | |||

Stk11
Genomic Map