siRNA Details
| Gene Symbol: Stk11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB015801.1 | - | 7 | 933 | ||
| AF129870.1 | - | 7 | 1311 | |||
| AF145287.1 | - | 7 | 915 | |||
| AF151711.1 | - | 7 | 915 | |||
| AK004499.1 | - | 5 | 594 | |||
| AK147802.1 | - | 7 | 975 | |||
| AK171909.1 | - | 8 | 1709 | |||
| AK172385.1 | - | 7 | 1698 | |||
| AK172528.1 | - | 7 | 1695 | |||
| BC052379.1 | - | 7 | 976 | 5688695 | ||
| EU730638.1 | - | 7 | 894 | |||

Stk11
Genomic Map