siRNA Details
| Gene Symbol: Tmem63a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144794.2 | NP_659043.1 | 11 | 977 | ||
| GenBank mRNA | AK049360.1 | - | 11 | 971 | ||
| AK144899.1 | - | 11 | 1030 | |||
| AK170559.1 | - | 11 | 993 | |||
| AK173027.1 | - | 11 | 970 | |||
| BC014795.1 | - | 11 | 950 | 3961992 | ||
| BC019442.1 | - | 11 | 977 | 4036849 | ||

Tmem63a
Genomic Map