siRNA Details
Gene Symbol: Dock3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153413.2 | NP_700462.2 | 12 | 1302 | ||
GenBank mRNA | AY145302.1 | - | 12 | 1252 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dock3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153413.2 | NP_700462.2 | 12 | 1302 | ||
GenBank mRNA | AY145302.1 | - | 12 | 1252 |