siRNA Details
| Gene Symbol: Adamts18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172466.2 | NP_766054.2 | 13 | 2269 | ||
| GenBank mRNA | AK036137.1 | - | 10 | 1348 | ||
| AK053851.1 | - | 12 | 2381 | |||
| AK140876.1 | - | 13 | 2045 | |||
| BC094674.1 | - | 13 | 1924 | 30548647 | ||
| BC144816.1 | - | 11 | 1527 | |||
| BC144817.1 | - | 11 | 1530 | |||

Adamts18
Genomic Map