siRNA Details
| Gene Symbol: Myo5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081322.1 | NP_001074791.1 | 40, 41 | 5209 | ||
| GenBank mRNA | AK033571.1 | - | 16, 17 | 2014 | ||
| AK052785.1 | - | 2, 3 | 1071 | |||
| AK075657.1 | - | 5, 6 | 717 | |||
| BC003985.1 | - | 11, 12 | 1665 | |||

Myo5c
Genomic Map