siRNA Details
| Gene Symbol: Gm595 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085499.1 | NP_001078968.1 | 1 | 157 | ||
| GenBank mRNA | AK133392.1 | - | 1 | 158 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm595| Gene Symbol: Gm595 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085499.1 | NP_001078968.1 | 1 | 157 | ||
| GenBank mRNA | AK133392.1 | - | 1 | 158 | ||