siRNA Details
| Gene Symbol: Gipc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148951.1 | NP_683753.1 | 4 | 592 | ||
| GenBank mRNA | AB074494.1 | - | 4 | 592 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gipc3| Gene Symbol: Gipc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148951.1 | NP_683753.1 | 4 | 592 | ||
| GenBank mRNA | AB074494.1 | - | 4 | 592 | ||