siRNA Details
| Gene Symbol: Stxbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011505.2 | NP_035635.1 | 16 | 1673 | ||
| GenBank mRNA | AF152924.1 | - | 16 | 1663 | ||
| AK039029.1 | - | 4 | 347 | |||
| AK138499.1 | - | 4 | 346 | |||
| BC032881.1 | - | 16 | 1627 | 1224878 | ||

Stxbp4
Genomic Map