siRNA Details
| Gene Symbol: Gan | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081151.1 | NP_001074620.1 | 9 | 1471 | ||
| GenBank mRNA | AK039455.1 | - | 9 | 1470 | ||
| BC112432.1 | - | 9 | 1470 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gan| Gene Symbol: Gan | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081151.1 | NP_001074620.1 | 9 | 1471 | ||
| GenBank mRNA | AK039455.1 | - | 9 | 1470 | ||
| BC112432.1 | - | 9 | 1470 | |||