siRNA Details
| Gene Symbol: Gpkow | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173747.3 | NP_776108.2 | 11 | 1499 | ||
| GenBank mRNA | AK028433.1 | - | 11 | 1421 | ||
| AK144167.1 | - | 11 | 1430 | |||
| BC062646.1 | - | 9 | 1049 | |||
| BC092224.1 | - | 11 | 1405 | 30355916 | ||
| BC120679.1 | - | 10 | 1182 | 40129602 | ||
| BC125335.1 | - | 10 | 1182 | 40129850 | ||

Gpkow
Genomic Map