siRNA Details
| Gene Symbol: Gpkow | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173747.3 | NP_776108.2 | 5, 6 | 870 | ||
| GenBank mRNA | AK028433.1 | - | 5, 6 | 792 | ||
| AK144167.1 | - | 5, 6 | 800 | |||
| BC062646.1 | - | 3, 4 | 420 | |||
| BC092224.1 | - | 5, 6 | 776 | 30355916 | ||
| BC120679.1 | - | 4, 5 | 553 | 40129602 | ||
| BC125335.1 | - | 4, 5 | 553 | 40129850 | ||

Gpkow
Genomic Map