siRNA Details
| Gene Symbol: Sectm1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145373.2 | NP_663348.2 | 2 | 195 | ||
| GenBank mRNA | AK082020.1 | - | 2 | 196 | ||
| AK170159.1 | - | 2 | 191 | |||
| AK171246.1 | - | 2 | 197 | |||
| AW985540.1 | - | 2 | 195 | |||
| BC010462.1 | - | 2 | 162 | 3585040 | ||

Sectm1a
Genomic Map