siRNA Details
| Gene Symbol: Frmd4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177843.1 | NP_001171314.1 | 21 | 3070 | ||
| NM_001177844.1 | NP_001171315.1 | 10 | 2198 | |||
| NM_172475.3 | NP_766063.3 | 21 | 2700 | |||
| GenBank mRNA | AB093292.1 | - | 7 | 1394 | ||
| AK082365.1 | - | 21 | 2646 | |||
| AK148414.1 | - | 10 | 2197 | |||
| AK171532.1 | - | 10 | 2126 | |||
| BC058672.1 | - | 21 | 2691 | 6830456 | ||
| BC082808.1 | - | 21 | 3077 | |||

Frmd4a
Genomic Map