siRNA Details
| Gene Symbol: Frmd4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177843.1 | NP_001171314.1 | 19 | 2382 | ||
| NM_001177844.1 | NP_001171315.1 | 8 | 1510 | |||
| NM_172475.3 | NP_766063.3 | 19 | 2012 | |||
| GenBank mRNA | AB093292.1 | - | 5 | 706 | ||
| AK082365.1 | - | 19 | 1958 | |||
| AK148414.1 | - | 8 | 1509 | |||
| AK171532.1 | - | 8 | 1438 | |||
| AK206319.1 | - | 3 | 255 | |||
| BC058672.1 | - | 19 | 2003 | 6830456 | ||
| BC082808.1 | - | 19 | 2389 | |||

Frmd4a
Genomic Map