siRNA Details
| Gene Symbol: Topaz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199736.1 | NP_001186665.1 | 20 | 4891 | ||
| GenBank mRNA | HM631980.1 | - | 20 | 4891 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Topaz1| Gene Symbol: Topaz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199736.1 | NP_001186665.1 | 20 | 4891 | ||
| GenBank mRNA | HM631980.1 | - | 20 | 4891 | ||