siRNA Details
| Gene Symbol: Tmc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172476.4 | NP_766064.2 | 7 | 920 | ||
| GenBank mRNA | AK082562.1 | - | 7 | 949 | ||
| AK083189.1 | - | 7 | 923 | |||
| AK132820.1 | - | 7 | 949 | |||
| AY236499.1 | - | 7 | 981 | |||
| AY263161.1 | - | 7 | 949 | |||

Tmc7
Genomic Map