siRNA Details
| Gene Symbol: Tmc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172476.4 | NP_766064.2 | 11 | 1486 | ||
| GenBank mRNA | AK082562.1 | - | 11 | 1515 | ||
| AK083189.1 | - | 11 | 1489 | |||
| AK132820.1 | - | 11 | 1516 | |||
| AY236499.1 | - | 11 | 1547 | |||
| AY263161.1 | - | 11 | 1515 | |||
| BC105966.1 | - | 5 | 517 | |||

Tmc7
Genomic Map