siRNA Details
| Gene Symbol: Tbc1d25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166437.1 | NP_001159909.1 | 5 | 542 | ||
| NM_172478.3 | NP_766066.2 | 4 | 749 | |||
| GenBank mRNA | AK040932.1 | - | 4 | 492 | ||
| AK078145.1 | - | 4 | 750 | |||
| AK166256.1 | - | 4 | 488 | |||
| BC060178.1 | - | 4 | 526 | |||
| BC113777.1 | - | 5 | 490 | |||
| BC128565.1 | - | 5 | 492 | |||
| BC128566.1 | - | 4 | 699 | 40094900 | ||

Tbc1d25
Genomic Map