siRNA Details
| Gene Symbol: Tbc1d25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166437.1 | NP_001159909.1 | 4 | 417 | ||
| NM_172478.3 | NP_766066.2 | 3 | 624 | |||
| GenBank mRNA | AK040932.1 | - | 3 | 367 | ||
| AK078145.1 | - | 3 | 625 | |||
| AK166256.1 | - | 3 | 363 | |||
| BC060178.1 | - | 3 | 401 | |||
| BC113777.1 | - | 4 | 365 | |||
| BC128565.1 | - | 4 | 367 | |||
| BC128566.1 | - | 3 | 574 | 40094900 | ||

Tbc1d25
Genomic Map