siRNA Details
| Gene Symbol: T | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009309.2 | NP_033335.1 | 6, 7 | 1003 | ||
| GenBank mRNA | X51683.1 | - | 6, 7 | 1003 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

T| Gene Symbol: T | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009309.2 | NP_033335.1 | 6, 7 | 1003 | ||
| GenBank mRNA | X51683.1 | - | 6, 7 | 1003 | ||