siRNA Details
| Gene Symbol: D130043K22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK051381.1 | - | 20 | 3299 | ||
| AK122246.1 | - | 21 | 3435 | |||
| BC115723.1 | - | 21 | 3354 | 40086618 | ||
| BC115940.1 | - | 21 | 3354 | 40086616 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D130043K22Rik| Gene Symbol: D130043K22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK051381.1 | - | 20 | 3299 | ||
| AK122246.1 | - | 21 | 3435 | |||
| BC115723.1 | - | 21 | 3354 | 40086618 | ||
| BC115940.1 | - | 21 | 3354 | 40086616 | ||