siRNA Details
| Gene Symbol: Slc30a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252478.1 | NP_001239407.1 | 5, 6 | 350 | ||
| NM_144798.6 | NP_659047.2 | 5, 6 | 335 | |||
| GenBank mRNA | AF233346.1 | - | 4, 5 | 287 | ||
| AF395840.1 | - | 4, 5 | 277 | |||
| AK082907.1 | - | 5, 6 | 312 | |||
| AK150342.1 | - | 5, 6 | 311 | |||
| AK151120.1 | - | 5, 6 | 311 | |||
| AK160903.1 | - | 5, 6 | 339 | |||
| BC066162.1 | - | 5, 6 | 302 | 30471027 | ||
| BC068169.1 | - | 5, 6 | 324 | 30542277 | ||

Slc30a6
Genomic Map