siRNA Details
| Gene Symbol: Dock10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285927.1 | NP_001272856.1 | 37 | 4218 | ||
| NM_175291.4 | NP_780500.2 | 37 | 4202 | |||
| GenBank mRNA | AK122347.1 | - | 28 | 3056 | ||
| BC150753.1 | - | 37 | 4138 | 9087664 | ||
| GU797481.1 | - | 37 | 4175 | |||

Dock10
Genomic Map