siRNA Details
| Gene Symbol: Dock10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285927.1 | NP_001272856.1 | 55 | 6509 | ||
| NM_175291.4 | NP_780500.2 | 55 | 6493 | |||
| GenBank mRNA | AK034064.1 | - | 11 | 1230 | ||
| AK122347.1 | - | 46 | 5254 | |||
| BC150753.1 | - | 55 | 6429 | 9087664 | ||
| BU698022.1 | - | 4 | 406 | |||
| GU797481.1 | - | 55 | 6466 | |||

Dock10
Genomic Map