siRNA Details
| Gene Symbol: Lrch2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081173.1 | NP_001074642.1 | 2, 3 | 547 | ||
| GenBank mRNA | AK144916.1 | - | 2, 3 | 377 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrch2| Gene Symbol: Lrch2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081173.1 | NP_001074642.1 | 2, 3 | 547 | ||
| GenBank mRNA | AK144916.1 | - | 2, 3 | 377 | ||