siRNA Details
| Gene Symbol: Mtmr9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177594.1 | NP_808262.1 | 6 | 1223 | ||
| GenBank mRNA | AK035026.1 | - | 6 | 1238 | ||
| AK044957.1 | - | 6 | 1253 | |||
| AK152320.1 | - | 6 | 1009 | |||
| BC046275.1 | - | 6 | 1223 | 6307469 | ||
| BC145946.1 | - | 6 | 978 | 40131249 | ||
| BC145948.1 | - | 6 | 978 | 40131251 | ||

Mtmr9
Genomic Map