siRNA Details
| Gene Symbol: Thsd7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172485.3 | NP_766073.2 | 15 | 3330 | ||
| GenBank mRNA | AK051714.1 | - | 15 | 3172 | ||
| AK083798.1 | - | 15 | 3158 | |||
| AK166575.1 | - | 3 | 514 | |||
| BC063250.1 | - | 15 | 3160 | 6410178 | ||

Thsd7b
Genomic Map