siRNA Details
| Gene Symbol: Tbc1d31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081396.2 | NP_001074865.2 | 15, 16 | 2388 | ||
| NM_001167679.1 | NP_001161151.1 | 15, 16 | 2388 | |||
| GenBank mRNA | AB185844.1 | - | 15, 16 | 2356 | ||
| AK141805.1 | - | 15, 16 | 2258 | |||
| AK154264.1 | - | 15, 16 | 2353 | |||
| AK171024.1 | - | 15, 16 | 2353 | |||
| BC050039.1 | - | 15, 16 | 2334 | 30016266 | ||
| BC066830.1 | - | 15, 16 | 2320 | 30463017 | ||

Tbc1d31
Genomic Map