siRNA Details
| Gene Symbol: Coq10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081040.1 | NP_001074509.1 | 4 | 1741 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CK346052.1 | - | 383 | Chr. - - [-] on Build | ||

Coq10a
Genomic Map