siRNA Details
| Gene Symbol: Pamr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173749.4 | NP_776110.3 | 9 | 1308 | ||
| GenBank mRNA | AK088017.1 | - | 9 | 1207 | ||
| BC031841.1 | - | 9 | 1262 | 3675412 | ||
| BC057685.1 | - | 9 | 1262 | 3675412 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pamr1| Gene Symbol: Pamr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173749.4 | NP_776110.3 | 9 | 1308 | ||
| GenBank mRNA | AK088017.1 | - | 9 | 1207 | ||
| BC031841.1 | - | 9 | 1262 | 3675412 | ||
| BC057685.1 | - | 9 | 1262 | 3675412 | ||