siRNA Details
| Gene Symbol: Gab3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181584.4 | NP_853615.1 | 2 | 451 | ||
| GenBank mRNA | AK031227.1 | - | 2 | 449 | ||
| AY057988.1 | - | 2 | 337 | |||
| BC141240.1 | - | 2 | 331 | 9055852 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gab3| Gene Symbol: Gab3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181584.4 | NP_853615.1 | 2 | 451 | ||
| GenBank mRNA | AK031227.1 | - | 2 | 449 | ||
| AY057988.1 | - | 2 | 337 | |||
| BC141240.1 | - | 2 | 331 | 9055852 | ||