siRNA Details
| Gene Symbol: Unc5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153135.3 | NP_694775.1 | 10 | 1537 | ||
| GenBank mRNA | AJ487854.1 | - | 10 | 1538 | ||
| BC137615.1 | - | 10 | 1328 | 8860634 | ||
| BC144783.1 | - | 8 | 1145 | 9053331 | ||
| BC144784.1 | - | 8 | 1145 | 9053332 | ||
| BC145695.1 | - | 7 | 1112 | 40130998 | ||
| FJ824810.1 | - | 9 | 1455 | |||

Unc5d
Genomic Map