siRNA Details
| Gene Symbol: Lacc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172488.2 | NP_766076.1 | 3, 4 | 918 | ||
| GenBank mRNA | AK033565.1 | - | 3, 4 | 919 | ||
| AK053693.1 | - | 3, 4 | 1203 | |||
| AK152154.1 | - | 3, 4 | 859 | |||
| AK153349.1 | - | 3, 4 | 943 | |||
| BC116748.1 | - | 2, 3 | 674 | 40126067 | ||
| BC119140.1 | - | 2, 3 | 674 | 8733889 | ||

Lacc1
Genomic Map