siRNA Details
| Gene Symbol: Gm382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033241.3 | NP_001028413.2 | 3 | 3075 | ||
| GenBank mRNA | AK132934.1 | - | 3 | 3076 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm382| Gene Symbol: Gm382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033241.3 | NP_001028413.2 | 3 | 3075 | ||
| GenBank mRNA | AK132934.1 | - | 3 | 3076 | ||