siRNA Details
Gene Symbol: Cpne9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170673.3 | NP_733773.2 | 18 | 1445 | ||
GenBank mRNA | AK042692.1 | - | 18 | 1446 | ||
AK044080.1 | - | 18 | 1411 | |||
AK047571.1 | - | 16 | 2460 | |||
AK137422.1 | - | 18 | 1446 | |||
AK220550.1 | - | 10 | 2809 | |||
BC115728.1 | - | 18 | 1395 | 40039295 | ||
BQ550678.1 | - | 2 | 80 |
