siRNA Details
| Gene Symbol: Mtss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144800.2 | NP_659049.2 | 12 | 1676 | ||
| GenBank mRNA | AK030533.1 | - | 10 | 1070 | ||
| AY214918.1 | - | 12 | 1177 | |||
| BC023682.1 | - | 6 | 782 | |||
| BC024131.1 | - | 12 | 1641 | 5101888 | ||

Mtss1
Genomic Map