siRNA Details
| Gene Symbol: Mtss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146180.1 | NP_001139652.1 | 14 | 2664 | ||
| NM_144800.2 | NP_659049.2 | 15 | 2706 | |||
| GenBank mRNA | AK030533.1 | - | 13 | 2100 | ||
| AK129139.1 | - | 14 | 2661 | |||
| AY214918.1 | - | 15 | 2207 | |||
| BC003483.1 | - | 1 | 625 | |||
| BC016555.1 | - | 4 | 1020 | |||
| BC023682.1 | - | 9 | 1812 | |||
| BC024131.1 | - | 15 | 2671 | 5101888 | ||
| BC042632.1 | - | 4 | 1025 | |||

Mtss1
Genomic Map