siRNA Details
| Gene Symbol: Exoc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177333.3 | NP_796307.2 | 3 | 277 | ||
| GenBank mRNA | AK088333.1 | - | 3 | 271 | ||
| AK089979.1 | - | 3 | 288 | |||
| AK131141.1 | - | 3 | 261 | |||
| AK141537.1 | - | 3 | 277 | |||
| BC031743.1 | - | 3 | 265 | 4187527 | ||

Exoc3
Genomic Map