siRNA Details
| Gene Symbol: Tmem87a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110496.1 | NP_001103966.1 | 13 | 1283 | ||
| NM_001110497.1 | NP_001103967.1 | 13 | 1283 | |||
| NM_173734.3 | NP_776095.2 | 13 | 1286 | |||
| GenBank mRNA | AK044591.1 | - | 13 | 1237 | ||
| AK086738.1 | - | 13 | 1233 | |||
| AK088559.1 | - | 13 | 1268 | |||
| AK134268.1 | - | 13 | 1249 | |||
| BC004047.1 | - | 13 | 1232 | |||
| BC024676.1 | - | 3 | 234 | |||
| BC027354.1 | - | 13 | 1259 | 4010477 | ||

Tmem87a
Genomic Map