siRNA Details
| Gene Symbol: Ccdc114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033243.2 | NP_001028415.1 | 13 | 2031 | ||
| GenBank mRNA | AK135860.1 | - | 13 | 2032 | ||
| BC013491.1 | - | 9 | 1351 | |||
| BC172013.1 | - | 11 | 2339 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc114| Gene Symbol: Ccdc114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033243.2 | NP_001028415.1 | 13 | 2031 | ||
| GenBank mRNA | AK135860.1 | - | 13 | 2032 | ||
| BC013491.1 | - | 9 | 1351 | |||
| BC172013.1 | - | 11 | 2339 | |||