siRNA Details
| Gene Symbol: Ptchd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001093750.1 | NP_001087219.1 | 3 | 2262 | ||
| GenBank mRNA | AK036111.1 | - | 3 | 2057 | ||
| BC116312.1 | - | 3 | 2260 | 40095445 | ||
| BC116313.1 | - | 3 | 2257 | 40095446 | ||

Ptchd1
Genomic Map