siRNA Details
Gene Symbol: Fancd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033244.3 | NP_001028416.2 | 15, 16 | 1367 | ||
GenBank mRNA | AK142384.1 | - | 15, 16 | 1365 | ||
AK155602.1 | - | 15, 16 | 1346 | |||
AK166411.1 | - | 15, 16 | 1337 | |||
BC040776.1 | - | 3, 4 | 248 | |||
BC042619.1 | - | 3, 4 | 248 | |||
BC139004.1 | - | 15, 16 | 1365 | 8862026 |
