siRNA Details
Gene Symbol: Fancd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033244.3 | NP_001028416.2 | 42, 43 | 4278 | ||
GenBank mRNA | AK142384.1 | - | 42, 43 | 4276 | ||
AK155602.1 | - | 41, 42 | 4218 | |||
BC040776.1 | - | 30, 31 | 3159 | |||
BC042619.1 | - | 30, 31 | 3159 | |||
BC139004.1 | - | 42, 43 | 4276 | 8862026 |
