siRNA Details
| Gene Symbol: Mfsd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172499.2 | NP_766087.1 | 4 | 387 | ||
| GenBank mRNA | AK029874.1 | - | 4 | 388 | ||
| AK169911.1 | - | 5 | 548 | |||
| AK170820.1 | - | 3 | 2050 | |||
| BC057633.1 | - | 4 | 393 | 4035998 | ||

Mfsd9
Genomic Map