siRNA Details
| Gene Symbol: Mfsd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172499.2 | NP_766087.1 | 2 | 243 | ||
| GenBank mRNA | AK029874.1 | - | 2 | 244 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mfsd9| Gene Symbol: Mfsd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172499.2 | NP_766087.1 | 2 | 243 | ||
| GenBank mRNA | AK029874.1 | - | 2 | 244 | ||