siRNA Details
| Gene Symbol: Ccdc73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177600.4 | NP_808268.2 | 16 | 2066 | ||
| GenBank mRNA | AK029862.1 | - | 8 | 1273 | ||
| BC095976.1 | - | 15 | 1812 | |||
| BC150667.1 | - | 7 | 1195 | 9087578 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc73| Gene Symbol: Ccdc73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177600.4 | NP_808268.2 | 16 | 2066 | ||
| GenBank mRNA | AK029862.1 | - | 8 | 1273 | ||
| BC095976.1 | - | 15 | 1812 | |||
| BC150667.1 | - | 7 | 1195 | 9087578 | ||