siRNA Details
| Gene Symbol: Pde12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178668.3 | NP_848783.3 | 1 | 1522 | ||
| GenBank mRNA | AK051991.1 | - | 738 | |||
| AK082947.1 | - | 1 | 1345 | |||
| AK088843.1 | - | 1 | 1335 | |||
| AK143567.1 | - | 1 | 1349 | |||
| AK155088.1 | - | 1356 | ||||
| AK161083.1 | - | 1 | 1562 | |||
| AK167708.1 | - | 1 | 1320 | |||
| AK201375.1 | - | 73 | ||||
| AK210569.1 | - | 1 | 73 | |||
| AK215000.1 | - | 1 | 73 | |||
| BC064450.1 | - | 1 | 1522 | 6417852 | ||

Pde12
Genomic Map